5-Benzyloxy-6-azaindole-3-carboxaldehyde 96%

Supplier: Thermo Scientific Chemicals
Warning

H50173-03 H50173-MD
AAAH50173-03EA 166 USD
AAAH50173-03 AAAH50173-MD
5-Benzyloxy-6-azaindole-3-carboxaldehyde 96%
5-Benzyloxy-6-azaindole-3-carboxaldehyde

Purity: 96% CAS No.: 56795-92-7 Molecular Formula: C15H12N2O2 Formula Weight: 252.28 MDL No.: MFCD00191784

Formula: C₁₅H₁₂N₂O₂
MW: 252.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191784
CAS Number: 56795-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR