5-Iodo-2-methoxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H50135-03 H50135-06
AAAH50135-03EA 125 USD
AAAH50135-03 AAAH50135-06
5-Iodo-2-methoxypyridine 98%
5-Iodo-2-methoxypyridine

Purity: 98% CAS No.: 13472-61-2 Molecular Formula: C6H6INO Formula Weight: 235.03 MDL No.: MFCD07781180

Formula: C₆H₆INO
Storage Temperature: Ambient
MDL Number: MFCD07781180
CAS Number: 13472-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR