2-Isopropoxypyridine-3-carboxaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

H50089-03 H50089-06
AAAH50089-03EA 114 USD
AAAH50089-03 AAAH50089-06
2-Isopropoxypyridine-3-carboxaldehyde 98%
2-Isopropoxypyridine-3-carboxaldehyde

Purity: 98% CAS No.: 885278-10-4 Molecular Formula: C9H11NO2 Formula Weight: 165.19 MDL No.: MFCD06804560

Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD06804560
CAS Number: 885278-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR