2-Ethoxypyridine-3-carboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Ethoxypyridine-3-carbaldehyde, 2-Ethoxynicotinaldehyde

H50088-03 H50088-06
AAAH50088-03EA 114 USD
AAAH50088-03 AAAH50088-06
2-Ethoxypyridine-3-carboxaldehyde 97%
2-Ethoxypyridine-3-carboxaldehyde

Purity: 97% CAS No.: 885278-07-9 Molecular Formula: C8H9NO2 Formula Weight: 151.17 MDL No.: MFCD06804559

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06804559
CAS Number: 885278-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR