3-(2-Methoxy-3-pyridyl)acrylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H50063-03 H50063-06
AAAH50063-03EA 136 USD
AAAH50063-03 AAAH50063-06
3-(2-Methoxy-3-pyridyl)acrylic acid 95%
3-(2-Methoxy-3-pyridyl)acrylic acid

Purity: 95% CAS No.: 131674-41-4 Molecular Formula: C9H9NO3 Formula Weight: 179.18 MDL No.: MFCD07781156

Formula: C₉H₉NO₃
MW: 179.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781156
CAS Number: 131674-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR