2-Chloro-4-methylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-4-methylbenzeneboronic acid

H28604-03 H28604-06
AAH28604-14EA 382.95 USD
AAH28604-14 AAAH28604-03 AAAH28604-06
2-Chloro-4-methylphenylboronic acid 97%
2-Chloro-4-methylphenylboronic acid

Purity: 97% CAS No.: 145349-62-8 Molecular Formula: C7H8BClO2 Formula Weight: 170.40 Melting Point: 198-202° MDL No.: MFCD03411936

Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD03411936
CAS Number: 145349-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR