6-Bromo-2,4-dimethylpyridine-3-boronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27460-03
H27460-MD
AAH27460-03EA
524
USD
AAH27460-03
AAH27460-MD
6-Bromo-2,4-dimethylpyridine-3-boronic acid 95%
6-Bromo-2,4-dimethylpyridine-3-boronic acid
Purity: 95%
CAS No.: 1072944-23-0
Molecular Formula: C7H9BBrNO2
Formula Weight: 229.87
Melting Point: ca 145° dec.
MDL No.: MFCD09037491
Formula:
C₇H₉BBrNO₂ MW: 229.87 g/mol Melting Pt: ca 145 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD09037491 CAS Number: 1072944-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...