(R,R)-(+)-2,3-Bis(diphenylphosphino)butane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2R,3R)-Butane-2,3-diylbis(diphenylphosphine), (2R,3R)-(+)-Bis(diphenylphosphino)butane
H27448-MD
H27448-03
H27448-MF
AAH27448-MDEA
242
USD
AAH27448-MD
AAH27448-03
AAH27448-MF
(R,R)-(+)-2,3-Bis(diphenylphosphino)butane 98%
(R,R)-(+)-2,3-Bis(diphenylphosphino)butane
Purity: 98%
CAS No.: 74839-84-2
Molecular Formula: C28H28P2
Formula Weight: 426.48
Melting Point: 104-109°
MDL No.: MFCD00075403
Formula:
C₂₈H₂₈P₂ Melting Pt: 104…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075403 CAS Number: 74839-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...