N,S-Dimethylisothiouronium hydriodide 98%

Supplier: Thermo Scientific Chemicals
Warning

H27439-06 H27439-14
AAH27439-06EA 37.6 USD
AAH27439-06 AAH27439-14
N,S-Dimethylisothiouronium hydriodide 98%
N,S-Dimethylisothiouronium hydriodide

Purity: 98% CAS No.: 41306-45-0 Molecular Formula: C3H8N2S.HI Formula Weight: 232.07 Melting Point: 134-136° MDL No.: MFCD00192092

Formula: C₃H₈N₂S·HI
MW: 232.07 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00192092
CAS Number: 41306-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR