6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, tert-Butyl methyl(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate
H27430-03
H27430-MD
AAH27430-03EA
273
USD
AAH27430-03
AAH27430-MD
6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester 95%
6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester
Purity: 95%
CAS No.: 1032758-87-4
Molecular Formula: C17H27BN2O4
Formula Weight: 334.22
Storage Temperature: 2°C to 8°C
Melting Point: 112-119°
MDL No.: MFCD09037484
Formula:
C₁₇H₂₇BN₂O₄ Melting Pt: 112…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037484 CAS Number: 1032758-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...