(+)-Bis[(R)-1-phenylethyl]amine ≥99%, ee 98%, ChiPros®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis[(R)-1-phenylethyl]amine

H27424-14 H27424-03 H27424-06
AAH27424-14EA 533 USD
AAH27424-14 AAH27424-03 AAH27424-06
(+)-Bis[(R)-1-phenylethyl]amine ≥99%, ee 98%, ChiPros®
(+)-Bis[(R)-1-phenylethyl]amine

Purity: 99% CAS No.: 23294-41-9 Molecular Formula: C16H19N Formula Weight: 225.34 Boiling Point: 98°/0.3mm MDL No.: MFCD00243088

Formula: C₁₆H₁₉N
Boiling Pt: 98 °C/0.3 mm
Storage Temperature: Ambient
MDL Number: MFCD00243088
CAS Number: 23294-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR