trans-4-Hydroxy-D-proline 97%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, H-D-Hyp-OH, H-trans-D-Hyp-OH

H27421-03 H27421-MD
AAH27421-03EA 233 USD
AAH27421-03 AAH27421-MD
trans-4-Hydroxy-D-proline 97%
trans-4-Hydroxy-D-proline

Purity: 97% CAS No.: 3398-22-9 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: ca 260° dec. MDL No.: MFCD00065608

Formula: C₅H₉NO₃
Melting Pt: ca 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00065608
CAS Number: 3398-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR