1-Methyl-4-piperidin-4-ylpiperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Methyl-4-(piperidin-4-yl)piperazine

H27408-03 H27408-06
AAH27408-03EA 66.8 USD
AAH27408-03 AAH27408-06
1-Methyl-4-piperidin-4-ylpiperazine 98%
1-Methyl-4-piperidin-4-ylpiperazine

Purity: 98% CAS No.: 53617-36-0 Molecular Formula: C10H21N3 Formula Weight: 183.30 Melting Point: 53-56° UN No.: 3259 MDL No.: MFCD03274729

Formula: C₁₀H₂₁N₃
MW: 183.3 g/mol
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD03274729
CAS Number: 53617-36-0
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR