2-Methoxy-4-nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

H27391-06 H27391-14
AAH27391-06EA 92.2 USD
AAH27391-06 AAH27391-14
2-Methoxy-4-nitrobenzonitrile 98%
2-Methoxy-4-nitrobenzonitrile

Purity: 98% CAS No.: 101084-96-2 Molecular Formula: C8H6N2O3 Formula Weight: 178.15 Melting Point: 177-180° UN No.: 3439 MDL No.: MFCD02093781

Formula: C₈H₆N₂O₃
MW: 178.15 g/mol
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD02093781
CAS Number: 101084-96-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR