4-Benzyloxy-6-chloropyrimidine 95%

Supplier: Thermo Scientific Chemicals
Warning

H27390-03 H27390-MD
AAH27390-03EA 316 USD
AAH27390-03 AAH27390-MD
4-Benzyloxy-6-chloropyrimidine 95%
4-Benzyloxy-6-chloropyrimidine

Purity: 95% CAS No.: 405930-65-6 Molecular Formula: C11H9ClN2O Formula Weight: 220.66 MDL No.: MFCD06798234

Formula: C₁₁H₉ClN₂O
MW: 220.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798234
CAS Number: 405930-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR