2-Ethyl-3,5(or 6)-dimethylpyrazine (mixture of isomers) 99%

Supplier: Thermo Scientific Chemicals
Warning

H27383-14 H27383-06
AAH27383-22EA 395.23 USD
AAH27383-22 AAH27383-14 AAH27383-06
2-Ethyl-3,5(or 6)-dimethylpyrazine (mixture of isomers) 99%
2-Ethyl-3,5(or 6)-dimethylpyrazine (mixture of isomers)

Purity: 99% CAS No.: 13925-07-0 Molecular Formula: C8H12N2 Formula Weight: 136.19 Flash Point: 66°(151°F) Refractive Index: 1.5015 Boiling Point: 182° MDL No.: MFCD00047392

Formula: C₈H₁₂N₂
MW: 136.194 g/mol
Boiling Pt: 182 °C
Density: 0.96
Flash Pt: 66 °C(151 °F)
Storage Temperature: Ambient
MDL Number: MFCD00047392
CAS Number: 27043-05-6
EINECS: 237-694-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR