Di-tert-butyl-N,N-diethylphosphoramidite 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Di-tert-butyl diethylphosphoramidite
H27380-03
H27380-06
AAH27380-03EA
89.6
USD
AAH27380-03
AAH27380-06
Di-tert-butyl-N,N-diethylphosphoramidite 94%
Di-tert-butyl-N,N-diethylphosphoramidite
Purity: 94%
CAS No.: 117924-33-1
Molecular Formula: [(CH3)3CO]2PN(CH2CH3)2
Formula Weight: 249.33
Storage Temperature: 2°C to 8°C
Flash Point: 78°(172°F)
Refractive Index: 1.434
Boiling Point: 39-41°/0.3mm
MDL No.: MFCD00134307
Formula:
C₁₂H₂₈NO₂P MW: 249.335 g/mol Boiling Pt: 39…41 °C/0.3 mm Density: 0.896 Flash Pt: 78 °C(172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00134307 CAS Number: 117924-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...