N-Boc-4-oxo-L-proline-tert-butyl ester 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Bis(tert-butyl) (2S)-4-oxopyrrolidine-1,2-dicarboxylate, (S)-Di-tert-butyl 4-oxopyrrolidine-1,2-dicarboxylate, Boc-4-Oxo-Pro-OtBu
H27375-06
H27375-03
AAH27375-06EA
667
USD
AAH27375-06
AAH27375-03
AAH27375-MD
N-Boc-4-oxo-L-proline-tert-butyl ester 97%
N-Boc-4-oxo-L-proline-tert-butyl ester
Purity: 97%
CAS No.: 166410-05-5
Molecular Formula: C14H23NO5
Formula Weight: 285.35
Storage Temperature: 2°C to 8°C
Melting Point: 64-67°
MDL No.: MFCD06797013
Formula:
C₁₄H₂₃NO₅ MW: 285.35 g/mol Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797013 CAS Number: 166410-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...