N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Cys(Trt)-OH, N-Fmoc-S-trityl-L-cysteine
H27363-06
H27363-14
AAH27363-06EA
64.36
USD
AAH27363-06
AAH27363-03
AAH27363-14
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine 95%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine
Purity: 95%
CAS No.: 103213-32-7
Molecular Formula: C37H31NO4S
Formula Weight: 585.71
Storage Temperature: 2°C to 8°C
Melting Point: 171-173°
Optical Rotation: +17° (c=1 in THF)
MDL No.: MFCD00038538
Formula:
C₃₇H₃₁NO₄S MW: 585.72 g/mol Melting Pt: 171…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038538 CAS Number: 103213-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...