2-Chloro-D-phenylalanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-D-Phe(2-Cl)-OH
H27359-03
H27359-MD
AAH27359-03EA
147
USD
AAH27359-03
AAH27359-MD
2-Chloro-D-phenylalanine 98%
2-Chloro-D-phenylalanine
Purity: 98%
CAS No.: 80126-50-7
Molecular Formula: C9H10ClNO2
Formula Weight: 199.64
Melting Point: 211-213°
MDL No.: MFCD00077920
Formula:
C₉H₁₀ClNO₂ MW: 199.64 g/mol Melting Pt: 211…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077920 CAS Number: 80126-50-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...