(R)-(+)-1-Boc-3-methylpiperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3R)-3-Methylpiperazine, N1-BOC protected, (R)-1-Boc-3-Methylpiperazine, (R)-1-Boc-3-methyl-piperazine

H27350-03 H27350-06
AAH27350-03EA 35.6 USD
AAH27350-03 AAH27350-06
(R)-(+)-1-Boc-3-methylpiperazine 97%
(R)-(+)-1-Boc-3-methylpiperazine

Purity: 97% CAS No.: 163765-44-4 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Melting Point: 40-45° Optical Rotation: +16° (c=1 in dioxane)

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 40…45 °C
Storage Temperature: Ambient
MDL Number: MFCD02683205
CAS Number: 163765-44-4

Order Now

Specification Test Results

Optical rotation +16° (c=1 in dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR