5'-Bromo-2'-hydroxy-3'-nitroacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(5-Bromo-2-hydroxy-3-nitrophenyl)ethanone

H27331-14 H27331-06
AAH27331-14EA 118 USD
AAH27331-14 AAH27331-06
5'-Bromo-2'-hydroxy-3'-nitroacetophenone 97%
5'-Bromo-2'-hydroxy-3'-nitroacetophenone

Purity: 97% CAS No.: 70978-54-0 Molecular Formula: C8H6BrNO4 Formula Weight: 260.05 Melting Point: 129-132° MDL No.: MFCD01631134

Formula: C₈H₆BrNO₄
MW: 260.05 g/mol
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD01631134
CAS Number: 70978-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR