3-Maleimidopropionic acid N-hydroxysuccinimide ester 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(3-Maleimidopropionyloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoate, N-Succinimidyl 3-Maleimidopropionat, 3-Maleimidopropionic Acid N-Succinimidyl Ester, N-Succinimidyl 3-maleimidopropionate, 3-(Maleimido)propionic acid N-hydroxysuccinimide ester, N-β-maleimidopropyl-oxysuccinimide ester, BMPS
H27329-03
H27329-MD
AAH27329-03EA
580.83
USD
AAH27329-03
AAH27329-MD
3-Maleimidopropionic acid N-hydroxysuccinimide ester 99%
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)
Purity: 99%
CAS No.: 55750-62-4
Molecular Formula: C11H10N2O6
Formula Weight: 266.21
Storage Temperature: 2°C to 8°C
Melting Point: 168-170°
MDL No.: MFCD00043141
Formula:
C₁₁H₁₀N₂O₆ MW: 266.21 g/mol Melting Pt: 168…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043141 CAS Number: 55750-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...