Methyl-4-oxopiperidine-3-carboxylate hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-oxopiperidine-3-carboxylate hydrochloride, Methyl 4-Oxopiperidine-3-carboxylate HCl

H27318-14 H27318-06
AAH27318-03EA 43.38 USD
AAH27318-03 AAH27318-14 AAH27318-06
Methyl-4-oxopiperidine-3-carboxylate hydrochloride 95%
Methyl-4-oxopiperidine-3-carboxylate hydrochloride

Purity: 95% CAS No.: 71486-53-8 Molecular Formula: C7H11NO3.HCl Formula Weight: 193.63 Melting Point: ca 175° dec. Beilstein Reference: 4273241 MDL No.: MFCD00012769

Formula: C₇H₁₂ClNO₃
MW: 193.63 g/mol
Melting Pt: ∼ 175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012769
CAS Number: 71486-53-8
EINECS: 275-526-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR