(R)-1-(Boc-amino)-2-propanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-tert-Butyl (2-hydroxypropyl)carbamate

H27296-03 H27296-06
AAH27296-03EA 88.5 USD
AAH27296-03 AAH27296-MD AAH27296-06
(R)-1-(Boc-amino)-2-propanol 98%
(R)-1-(Boc-amino)-2-propanol

Purity: 98% CAS No.: 119768-44-4 Molecular Formula: C8H17NO3 Formula Weight: 175.23 Refractive Index: 1.4460 Optical Rotation: -24° (c=1 in chloroform) MDL No.: MFCD04974338

Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974338
CAS Number: 119768-44-4

Order Now

Specification Test Results

Optical rotation -24° (c=1 in chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR