(S)-2-Amino-4-phenylbutanoic acid 98%
Supplier: Thermo Scientific Chemicals
H27295-06
H27295-14
H27295-03
AAH27295-06EA
198
USD
AAH27295-06
AAH27295-14
AAH27295-03
(S)-2-Amino-4-phenylbutanoic acid 98%
(S)-2-Amino-4-phenylbutanoic acid
Purity: 98%
CAS No.: 943-73-7
Molecular Formula: C10H13NO2
Formula Weight: 179.22
Melting Point:
Formula:
C₁₀H₁₃NO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002619 CAS Number: 943-73-7 EINECS: 213-403-3 |
Specification Test Results
Optical rotation | +45° (c=1 in 3N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...