1,1'-Bis(dichlorophosphino)ferrocene 98%

Supplier: Thermo Scientific Chemicals
Danger

H27293-03 H27293-MD
AAH27293-03EA 155 USD
AAH27293-03 AAH27293-MD
1,1'-Bis(dichlorophosphino)ferrocene 98%
1,1'-Bis(dichlorophosphino)ferrocene

Purity: 98% CAS No.: 142691-70-1 Molecular Formula: C10H8Cl4FeP2 Formula Weight: 387.78 UN No.: 3261 MDL No.: MFCD01630854

Formula: C₁₀H₈Cl₄FeP₂
MW: 387.77 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01630854
CAS Number: 142691-70-1
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR