2-Chloropyridine-N-oxide hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H27280-06 H27280-14
AAH27280-06EA 75.1 USD
AAH27280-06 AAH27280-14
2-Chloropyridine-N-oxide hydrochloride 97%
2-Chloropyridine-N-oxide hydrochloride

Purity: 97% CAS No.: 20295-64-1 Molecular Formula: C5H4ClNO.HCl Formula Weight: 166.01 Melting Point: 138-142° MDL No.: MFCD00007913

Formula: C₅H₄ClNO·HCl
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00007913
CAS Number: 20295-64-1
EINECS: 243-712-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR