L-m-Tyrosine ≥97%

Supplier: Thermo Scientific Chemicals

H27224-03 H27224-MD
AAH27224-03EA 390 USD
AAH27224-03 AAH27224-MD
L-m-Tyrosine ≥97%
L-m-Tyrosine

Purity: 97+% CAS No.: 587-33-7 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Melting Point: 260-270° MDL No.: MFCD00063059

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Melting Pt: 260…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00063059
CAS Number: 587-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR