2-Hydroxycyclohexanone dimer 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27199-03
H27199-06
H27199-14
AAH27199-03EA
26.5
USD
AAH27199-03
AAH27199-06
AAH27199-14
2-Hydroxycyclohexanone dimer 90+%
2-Hydroxycyclohexanone dimer
Purity: 90+%
CAS No.: 30282-14-5
Molecular Formula: C12H20O4
Formula Weight: 228.28
Flash Point: 79°(174°F)
Melting Point: 100-107°
Boiling Point: 83-86°/13mm
UN No.: 1325
MDL No.: MFCD00151157
Formula:
C₆H₁₀O₂ MW: 228.28 g/mol Boiling Pt: 83…86 °C/13 mm Melting Pt: 100…107 °C Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00151157 CAS Number: 30282-14-5 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...