4-Methyl-2H-1,4-benzoxazin-3(4H)-one 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27196-06
H27196-14
AAH27196-06EA
29.9
USD
AAH27196-06
AAH27196-14
4-Methyl-2H-1,4-benzoxazin-3(4H)-one 98%
4-Methyl-2H-1,4-benzoxazin-3(4H)-one
Purity: 98%
CAS No.: 21744-84-3
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Melting Point: 58-60°
MDL No.: MFCD00601365
Formula:
C₉H₉NO₂ Melting Pt: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00601365 CAS Number: 21744-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...