1-O-Methyl-2-deoxy-D-ribose 96%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,3S)-2-(Hydroxymethyl)-5-methoxytetrahydrofuran-3-ol

H27181-03 H27181-06
AAH27181-03EA 76.2 USD
AAH27181-03 AAH27181-06
1-O-Methyl-2-deoxy-D-ribose 96%
1-O-Methyl-2-deoxy-D-ribose

Purity: 96% CAS No.: 60134-26-1 Molecular Formula: C6H12O4 Formula Weight: 148.17 Flash Point: 110°(230°F) Refractive Index: 1.4690 Optical Rotation: +23° (c=1 in water) MDL No.: MFCD00134161

Formula: C₆H₁₂O₄
MW: 148.17 g/mol
Density: 1.224
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134161
CAS Number: 60134-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR