3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27175-06
H27175-03
AAH27175-06EA
121
USD
AAH27175-06
AAH27175-03
3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid 98%
3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid
Purity: 98%
CAS No.: 33632-74-5
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: 146-148°
MDL No.: MFCD00276310
Formula:
C₁₀H₁₀O₄ Melting Pt: 146…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00276310 CAS Number: 20825-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...