3-Bromomethylquinoxaline-2-one 90%, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
H27169-06
H27169-14
AAH27169-06EA
34.7
USD
AAH27169-06
AAH27169-14
3-Bromomethylquinoxaline-2-one 90%, tech.
3-Bromomethylquinoxaline-2-one
Purity: 90%
CAS No.: 62235-61-4
Molecular Formula: C9H7BrN2O
Formula Weight: 239.07
Melting Point:
Formula:
C₉H₇BrN₂O MW: 239.07 g/mol Melting Pt: >300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03412060 CAS Number: 62235-61-4 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...