5-Fluoro-1,3-dimethyl-2-nitrobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27166-03
H27166-06
AAH27166-03EA
79.2
USD
AAH27166-03
AAH27166-06
5-Fluoro-1,3-dimethyl-2-nitrobenzene 98%
5-Fluoro-1,3-dimethyl-2-nitrobenzene
Purity: 98%
CAS No.: 315-12-8
Molecular Formula: C8H8FNO2
Formula Weight: 169.16
Melting Point: 52-57°
MDL No.: MFCD03413407
Formula:
C₈H₈FNO₂ MW: 169.16 g/mol Melting Pt: 52…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD03413407 CAS Number: 315-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...