(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Boc-D-Phenylglycinol, (-)-N-Boc-D-α-phenylglycinol
H27163-06
H27163-03
AAH27163-06EA
235
USD
AAH27163-06
AAH27163-03
AAH27163-MD
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol 99%
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol
Purity: 99%
CAS No.: 102089-74-7
Molecular Formula: C13H19NO3
Formula Weight: 237.30
Melting Point: 137-141°
Optical Rotation: -40° (c=1 in chloroform)
MDL No.: MFCD00274205
Formula:
C₁₃H₁₉NO₃ Melting Pt: 137…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274205 CAS Number: 102089-74-7 |
Specification Test Results
Optical rotation | -40° (c=1 in chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...