6-Acetyl-2(3H)-benzothiazolone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27154-03
H27154-09
AAH27154-03EA
30.8
USD
AAH27154-03
AAH27154-09
6-Acetyl-2(3H)-benzothiazolone 97%
6-Acetyl-2(3H)-benzothiazolone
Purity: 97%
CAS No.: 133044-44-7
Molecular Formula: C9H7NO2S
Formula Weight: 193.22
Melting Point: 190-194°
MDL No.: MFCD02660572
Formula:
C₉H₇NO₂S Melting Pt: 190…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD02660572 CAS Number: 133044-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...