2-Chloro-L-phenylalanine 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Phe(2-Cl)-OH
H27135-MD
H27135-03
AAH27135-MDEA
151
USD
AAH27135-MD
AAH27135-03
2-Chloro-L-phenylalanine 98+%
2-Chloro-L-phenylalanine
Purity: 98+%
CAS No.: 103616-89-3
Molecular Formula: C9H10ClNO2
Formula Weight: 199.64
Melting Point: 196-199°
MDL No.: MFCD00077921
Formula:
C₉H₁₀ClNO₂ MW: 199.6 g/mol Melting Pt: 196…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077921 CAS Number: 103616-89-3 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...