Triethylsulfonium bis(trifluoromethanesulfonyl)imide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27116-18
H27116-06
AAH27116-18EA
339
USD
AAH27116-18
AAH27116-06
Triethylsulfonium bis(trifluoromethanesulfonyl)imide 98%
Triethylsulfonium bis(trifluoromethanesulfonyl)imide
Purity: 98%
CAS No.: 321746-49-0
Molecular Formula: (CF3SO2)2NS(CH2CH3)3
Formula Weight: 399.39
MDL No.: MFCD08276438
Formula:
C₈H₁₅F₆NO₄S₃ MW: 399.38 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08276438 CAS Number: 321746-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...