(S)-2-Methylpropane-2-sulfinamide 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-tert-Butylsulphinamide, (S)-(-)-tert-Butylsulphinamide, (S)-(-)-t-Butylsulphinamide, (S)-(-)-2-Methyl-2-propanesulphinamide, (S)-(-)-tert-Butyl sulphinamide
H27115-14
H27115-03
H27115-06
AAH27115-14EA
905
USD
AAH27115-14
AAH27115-03
AAH27115-06
(S)-2-Methylpropane-2-sulfinamide 97%
(S)-2-Methylpropane-2-sulfinamide
Purity: 97%
CAS No.: 343338-28-3
Molecular Formula: C4H11NOS
Formula Weight: 121.20
Storage Temperature: 2°C to 8°C
Melting Point: 97-101°
MDL No.: MFCD05861480
Formula:
C₄H₁₁NOS MW: 121.20 g/mol Melting Pt: 97…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD05861480 CAS Number: 343338-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...