Boc-trans-4-hydroxy-L-proline 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Boc-Hyp-OH, trans-N-Boc-4-hydroxy-L-proline
H27110-06
H27110-14
AAH27110-03EA
34.25
USD
AAH27110-03
AAH27110-06
AAH27110-14
Boc-trans-4-hydroxy-L-proline 97%
Boc-trans-4-hydroxy-L-proline
Purity: 97%
CAS No.: 13726-69-7
Molecular Formula: C10H17NO5
Formula Weight: 231.25
Melting Point: 123-127°
MDL No.: MFCD00053370
Formula:
C₁₀H₁₇NO₅ Melting Pt: 123…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053370 CAS Number: 13726-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...