D-Cysteine hydrochloride monohydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, H-D-Cys-OH·HCl·H₂O, H-D-Cys-OH.HCl

H27107-03 H27107-06
AAH27107-03EA 101 USD
AAH27107-03 AAH27107-06
D-Cysteine hydrochloride monohydrate 99%
D-Cysteine hydrochloride monohydrate

Purity: 99% CAS No.: 207121-46-8 Molecular Formula: C3H7NO2S.HCl.H2O Formula Weight: 175.64 (157.62anhy) Storage Temperature: 2°C to 8°C Melting Point: 184-186° Optical Rotation: -5.5° (c=5 in 5N HCl) MDL No.: MFCD00150051

Formula: C₃H₇NO₂S·HCl·H₂O
MW: 175.64 g/mol (157.62 anhy)
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00150051
CAS Number: 207121-46-8
EINECS: 251-043-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR