(-)-Bis[(S)-1-phenylethyl]amine ≥99%, ee 98%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bis[(S)-1-phenylethyl]amine
H27106-03
H27106-06
H27106-14
AAH27106-03EA
52
USD
AAH27106-03
AAH27106-06
AAH27106-14
(-)-Bis[(S)-1-phenylethyl]amine ≥99%, ee 98%, ChiPros®
(-)-Bis[(S)-1-phenylethyl]amine
Purity: 99%
CAS No.: 56210-72-1
Molecular Formula: C16H19N
Formula Weight: 225.34
Boiling Point: 98°/0.3mm
MDL No.: MFCD00243087
Formula:
C₁₆H₁₉N MW: 225.34 Boiling Pt: 98 °C/0.3 mm Storage Temperature: Ambient |
MDL Number:
MFCD00243087 CAS Number: 56210-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...