Tetrakis(dimethylamino)silane 99%

Supplier: Thermo Scientific Chemicals
Danger

H27092-03 H27092-09
AAH27092-03EA 34 USD
AAH27092-03 AAH27092-09
Tetrakis(dimethylamino)silane 99%
Tetrakis(dimethylamino)silane

Purity: 99% CAS No.: 1624-01-7 Molecular Formula: C8H24N4Si Formula Weight: 204.40 Flash Point: 58°(136°F) Refractive Index: 1.4430 Odor: Amine-like Melting Point: 15° Boiling Point: 196° UN No.: 3129 MDL No.: MFCD00014859

Formula: C₈H₂₄N₄Si
Boiling Pt: 196 °C
Melting Pt: 15 °C
Density: 0.885
Flash Pt: 58 °C(136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014859
CAS Number: 1624-01-7
EINECS: 216-611-2
UN: 3129
ADR: "4.3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR