4-Bromo-4'-tert-butylbiphenyl 98%

Supplier: Thermo Scientific Chemicals
Warning

H27076-09 H27076-03
AAH27076-09EA 278 USD
AAH27076-09 AAH27076-03
4-Bromo-4'-tert-butylbiphenyl 98%
4-Bromo-4'-tert-butylbiphenyl

Purity: 98% CAS No.: 162258-89-1 Molecular Formula: C16H17Br Formula Weight: 289.21 Melting Point: 140-143° Boiling Point: 152°/2mm MDL No.: MFCD01321141

Formula: C₁₆H₁₇Br
MW: 289.21 g/mol
Boiling Pt: 152 °C/2 mm
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD01321141
CAS Number: 162258-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR