2-(Diphenylphosphino)ethylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

H27070-03 H27070-09
AAH27070-03EA 165.43 USD
AAH27070-03 AAH27070-09
2-(Diphenylphosphino)ethylamine 95%
2-(Diphenylphosphino)ethylamine

Purity: 95% CAS No.: 4848-43-5 Molecular Formula: (C6H5)2PCH2CH2NH2 Formula Weight: 229.27 UN No.: 2735 MDL No.: MFCD00233831

Formula: (C₆H₅)₂PCH₂CH₂NH₂
MW: 229.257 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233831
CAS Number: 4848-43-5
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR