N-Benzyloxycarbonyl-L-alaninol

Supplier: Thermo Scientific Chemicals

Synonyms: Z-Ala-ol, (S)-Benzyl (1-hydroxypropan-2-yl)carbamate, (S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl ester, N-Cbz-L-Alaninol

H27066-06 H27066-03
AAH27066-06EA 179 USD
AAH27066-06 AAH27066-03
N-Benzyloxycarbonyl-L-alaninol
N-Cbz-L-Alaninol

CAS No.: 66674-16-6 Molecular Formula: C11H15NO3 Formula Weight: 209.25 MDL No.: MFCD00672530

Formula: C₁₁H₁₅NO₃
MW: 209.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672530
CAS Number: 66674-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR