4-Methoxy-2-(trifluoromethyl)phenylboronic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methoxy-2-(trifluoromethyl)benzeneboronic acid, (4-Methoxy-2-(trifluoromethyl)phenyl)boronic acid
H27062-03
H27062-06
AAH27062-03EA
121
USD
AAH27062-03
AAH27062-06
4-Methoxy-2-(trifluoromethyl)phenylboronic acid
4-Methoxy-2-(trifluoromethyl)phenylboronic acid
CAS No.: 313546-16-6
Molecular Formula: C8H8BF3O3
Formula Weight: 219.95
Storage Temperature: 2°C to 8°C
Melting Point: 168-172°
MDL No.: MFCD03095346
Formula:
C₈H₈BF₃O₃ MW: 219.95 g/mol Melting Pt: 168…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095346 CAS Number: 313546-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...