Phenyl-β-D-galactopyranoside 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Phenyl b-D-galactoside
H27028-03
H27028-06
AAH27028-03EA
74.1
USD
AAH27028-03
AAH27028-06
Phenyl-β-D-galactopyranoside 99%
Phenyl-beta-D-galactopyranoside
Purity: 99%
CAS No.: 2818-58-8
Molecular Formula: C12H16O6
Formula Weight: 256.25
Storage Temperature: 2°C to 8°C
Melting Point: 152-156°
Beilstein Reference: 87518
Optical Rotation: -42° (c=2 in water)
MDL No.: MFCD00063258
Formula:
C₁₂H₁₆O₆ MW: 256.25 g/mol Melting Pt: 152…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063258 CAS Number: 2818-58-8 EINECS: 220-573-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...