tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-BOC-3-pyrrolidinone
H27023-03
H27023-09
AAH27023-03EA
58.8
USD
AAH27023-03
AAH27023-09
tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
tert-Butyl-3-oxopyrrolidine-1-carboxylate
Purity: 97%
CAS No.: 101385-93-7
Molecular Formula: C9H15NO3
Formula Weight: 185.22
Storage Temperature: 2°C to 8°C
Melting Point: 34-38°
MDL No.: MFCD01631194
Formula:
C₉H₁₅NO₃ MW: 185.22 g/mol Melting Pt: 34…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631194 CAS Number: 101385-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...